Please login to your online account to display your discounted pricing

1,3-Dichloroisoquinoline, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 1, 3-Dichloroisoquinoline
Assay Percent Range 97%
Molecular Formula C9H5Cl2N
Formula Weight 198.05
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC382410050

  SDS
thermo fisher scientific
AC382410050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC382410010

  SDS
thermo fisher scientific
AC382410010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 198.05
Quantity 5g
Assay Percent Range 97%
Molecular Formula C9H5Cl2N
Packaging Glass bottle
CAS 7742-73-6
Chemical Name or Material 1, 3-Dichloroisoquinoline