missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dichloroisoquinoline, 97%

$94.84 - $328.47

Chemical Identifiers

CAS 7742-73-6
Molecular Formula C9H5Cl2N
Molecular Weight (g/mol) 198.046
MDL Number MFCD00034750
InChI Key BRGZEQXWZWBPJH-UHFFFAOYSA-N
Synonym isoquinoline, 1,3-dichloro, 1,3-dichloro-isoquinoline, pubchem7254, 1, 3-dichloroisoquinoline, acmc-209p9x, ksc493q3b, 1,3-dichloroisoquinoline
PubChem CID 298625
IUPAC Name 1,3-dichloroisoquinoline
SMILES C1=CC=C2C(=C1)C=C(N=C2Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5987303 View Documents Alfa Aesar™
H5987303
1g Each for $94.84
 
AAH5987306 View Documents Alfa Aesar™
H5987306
5g Each for $328.47
 
Specifications

Chemical Identifiers

7742-73-6
198.046
BRGZEQXWZWBPJH-UHFFFAOYSA-N
298625
C1=CC=C2C(=C1)C=C(N=C2Cl)Cl
C9H5Cl2N
MFCD00034750
isoquinoline, 1,3-dichloro, 1,3-dichloro-isoquinoline, pubchem7254, 1, 3-dichloroisoquinoline, acmc-209p9x, ksc493q3b, 1,3-dichloroisoquinoline
1,3-dichloroisoquinoline

Specifications

121°C to 122°C
7742-73-6
MFCD00034750
BRGZEQXWZWBPJH-UHFFFAOYSA-N
1,3-dichloroisoquinoline
298625
97%
1g
C9H5Cl2N
isoquinoline, 1,3-dichloro, 1,3-dichloro-isoquinoline, pubchem7254, 1, 3-dichloroisoquinoline, acmc-209p9x, ksc493q3b, 1,3-dichloroisoquinoline
C1=CC=C2C(=C1)C=C(N=C2Cl)Cl
198.046
198.05
1,3-Dichloroisoquinoline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning