Please login to your online account to display your discounted pricing

1,3-Dichlorobutane 99%, ACROS Organics™


Quantity 50mL

Manufacturer: thermo fisher scientific  113250500

 View more versions of this product

Catalog No. AC113250500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 127.01
Assay ≥98.5%
Quantity 50mL
Color Clear colorless
EINECS Number 214-718-9
TSCA TSCA
Density 1.11g/mL
Beilstein 01, I, 38
CAS Max % 100.0
Infrared Spectrum Authentic
MDL Number MFCD00000870
Health Hazard 1 LACHRYMATOR
CAS Min % 98.5
Refractive Index 1.4421 to 1.4441 (20°C, 589nm)
Chemical Name or Material 1, 3-Dichlorobutane
Assay Percent Range 99%
Name Note 99%
CAS 1190-22-3
Linear Formula CH3CH(Cl)CH2CH2Cl
Molecular Formula C4H8Cl2

Specifications

Chemical Name or Material 1, 3-Dichlorobutane
Name Note 99%
CAS 1190-22-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C4H8Cl2
Linear Formula CH3CH(Cl)CH2CH2Cl
Formula Weight 127.01
MDL Number MFCD00000870
Accessories

LACHRYMATOR

SDS & Safety Documents