Please login to your online account to display your discounted pricing

1,3-Dichlorobutane 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 3-Dichlorobutane
Name Note 99%
CAS 1190-22-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C4H8Cl2
Linear Formula CH3CH(Cl)CH2CH2Cl
Formula Weight 127.01
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC113250500

  SDS
thermo fisher scientific
113250500
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 127.01
Assay ≥98.5%
Quantity 50mL
Color Clear colorless
EINECS Number 214-718-9
TSCA TSCA
Density 1.11g/mL
Beilstein 01, I, 38
CAS Max % 100.0
Infrared Spectrum Authentic
MDL Number MFCD00000870
Health Hazard 1 LACHRYMATOR
CAS Min % 98.5
Refractive Index 1.4421 to 1.4441 (20°C, 589nm)
Chemical Name or Material 1, 3-Dichlorobutane
Assay Percent Range 99%
Name Note 99%
CAS 1190-22-3
Linear Formula CH3CH(Cl)CH2CH2Cl
Molecular Formula C4H8Cl2
Safety and Handling

LACHRYMATOR