Learn More
1,3-Dichloroacetone, typically 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 02230222
Description
1,3-Dichloroacetone was used as acceptor substrate in the cross-aldol reaction with donor substrates such as acetone, cyclopentanone and cyclohexanon
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,3-Dichloroacetone | |
43°C to 45°C | |
173°C | |
Odorless | |
1.44 | |
100 g | |
605456 | |
1,3-dichloroacetone, 1,3-dichloro-2-propanone, 1,3-dichloropropanone, s-dichloroacetone, sym-dichloroacetone, bis chloromethyl ketone, 2-propanone, 1,3-dichloro, acetone, 1,3-dichloro, chloromethyl ketone | |
SUNMBRGCANLOEG-UHFFFAOYSA-N | |
1,3-dichloropropan-2-one | |
10793 | |
99% (typically) |
534-07-6 | |
1.3834 g/mL at 46°C | |
89°C (193°F) | |
C3H4Cl2O | |
MFCD00000937 | |
UN2649 | |
14,3052 | |
Soluble in alcohol,diethyl ether,water. | |
ClCC(=O)CCl | |
126.96 | |
126.97 | |
Crystalline |
Chemical Identifiers
534-07-6 | |
126.96 | |
SUNMBRGCANLOEG-UHFFFAOYSA-N | |
10793 | |
ClCC(=O)CCl |
C3H4Cl2O | |
MFCD00000937 | |
1,3-dichloroacetone, 1,3-dichloro-2-propanone, 1,3-dichloropropanone, s-dichloroacetone, sym-dichloroacetone, bis chloromethyl ketone, 2-propanone, 1,3-dichloro, acetone, 1,3-dichloro, chloromethyl ketone | |
1,3-dichloropropan-2-one |
Safety and Handling
GHS H Statement
H300-H310-H330-H314-H318-H341
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
P201-P202-P260-P262-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H300+H310+H330-H314-H335-H340
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 1,3-DICHLOROACETONE
EINECSNumber : 208-585-6
RTECSNumber : UC1430000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only