Please login to your online account to display your discounted pricing

1,3-Dichloro-5,5-dimethylhydantoin, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 3-Dichloro-5, 5-dimethylhydantoin
Assay ≥66% (available chlorine)
Molecular Formula C5H6Cl2N2O2
Formula Weight 197.02
MDL Number MFCD00003190
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC214220050

  SDS
thermo fisher scientific
214220050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 197.02
Assay ≥66% (available chlorine)
Physical Form Crystalline Powder
Quantity 5g
Color White
RTECS Number MU0700000
TSCA TSCA
EINECS Number 204-258-7
Sublimation Point 100.0 °C
Melting Point 131° to 136°C
Beilstein 24, II, 158
Merck Index 15, 3074
Health Hazard 1 Danger
Infrared Spectrum Authentic
Packaging Glass bottle
Volatiles 0.5% max.
Insoluble Matter 1% max. (in chloroform)
Health Hazard 3 GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.
May cause an allergic skin reaction.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.
MDL Number MFCD00003190
Chemical Name or Material 1, 3-Dichloro-5, 5-dimethylhydantoin
Molecular Formula C5H6Cl2N2O2
CAS 118-52-5
Safety and Handling

Danger

GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.
May cause an allergic skin reaction.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.