Learn More
1,3-Dichloro-2-propanol, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 180180010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1, 3-Dichloro-2-propanol | |
96-23-1 | |
100.0 | |
172.0°C to 176.0°C | |
Authentic | |
Glass bottle | |
1.4820 to 1.4840 | |
MFCD00000951 | |
01, 364 | |
15, 3084 | |
Solubility in water: 13g/L (20°C). Other solubilities: miscible with most common organic solvents | |
OC(CCl)CCl | |
128.98 | |
13.9 mPa.s (20°C) | |
128.99 |
99% | |
98.5 | |
1.3510g/mL | |
74°C | |
98.5% min. (GC) | |
C3H6Cl2O | |
ClCH2CH(OH)CH2Cl | |
1L | |
1.351 | |
1,3-dichloro-2-propanol, dichlorohydrin, 1,3-dichloroisopropanol, 1,3-dichlorohydrin, glycerol 1,3-dichlorohydrin, 2-propanol, 1,3-dichloro, enodrin, 1,3-dichloro-2-hydroxypropane, 1,3-dichloroisopropyl alcohol, sym-glycerol dichlorohydrin | |
DEWLEGDTCGBNGU-UHFFFAOYSA-N | |
1,3-dichloropropan-2-ol | |
7289 | |
CHEBI:18917 | |
99% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
May cause cancer.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/fa
GHS Signal Word: Danger
EINECSNumber : 202-491-9
RTECSNumber : UB1400000
TSCA : TSCA
RUO â Research Use Only