Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dichloro-2-nitrosobenzene, 98%

$65.42 - $65.42

Chemical Identifiers

Chemical Name or Material 1,3-Dichloro-2-nitrosobenzene
CAS 1194-66-7
Molecular Formula C6H3Cl2NO
Formula Weight 176
MDL Number MFCD00051460
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2283001

 
alfa aesar™
B2283001
0.25g Each for $65.42
Description & Specifications

Specifications

Formula Weight 176
Chemical Name or Material 1,3-Dichloro-2-nitrosobenzene
Health Hazard 1 Warning
Assay 98%
Molecular Formula C6H3Cl2NO
MDL Number MFCD00051460
CAS 1194-66-7
Melting Point 170° to 173°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Quantity 0.25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C