missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dichloro-2-methoxy-5-nitrobenzene, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 27102-0500

 View more versions of this product

Catalog No. AC271020500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
Crystals
Slightly beige
100.0
17742-69-7
Glass bottle
99%
1, 3-Dichloro-2-methoxy-5-nitrobenzene
222.03
50g
97° to 100°C
99%
98.5
Authentic
2, 6-Dichloro-4-nitroanisole
C7 H5 Cl2 N O3

Chemical Identifiers

17742-69-7
100.0
C7 H5 Cl2 N O3
2, 6-Dichloro-4-nitroanisole
≥98.5%
98.5
99%
222.03
1, 3-Dichloro-2-methoxy-5-nitrobenzene, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger