missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dichloro-2-methoxy-5-nitrobenzene, 99%, ACROS Organics™

Chemical Identifiers

CAS 17742-69-7
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C7 H5 Cl2 N O3
Formula Weight 222.03
Synonym 2, 6-Dichloro-4-nitroanisole
Chemical Name or Material 1, 3-Dichloro-2-methoxy-5-nitrobenzene, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC271020500 SDS Thermo Fisher Scientific
27102-0500
Glass bottle 50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

17742-69-7
100.0
C7 H5 Cl2 N O3
2, 6-Dichloro-4-nitroanisole
≥98.5%
98.5
99%
222.03
1, 3-Dichloro-2-methoxy-5-nitrobenzene, 99%

Specifications

Slightly beige
50g
98.5
Authentic
99%
222.03
2, 6-Dichloro-4-nitroanisole
≥98.5%
97° to 100°C
17742-69-7
100.0
Crystals
C7 H5 Cl2 N O3
Glass bottle
1, 3-Dichloro-2-methoxy-5-nitrobenzene, 99%

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger