missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dichloro-2-iodobenzene, 98%

Manufacturer:  Alfa Aesar™ H5536514

 View more versions of this product

Catalog No. AAH5536514



Specifications

Specifications

1,3-Dichloro-2-iodobenzene
25g
C6H3Cl2I
#
C1=CC(=C(C(=C1)Cl)I)Cl
272.894
272.9
64°C to 67°C
19230-28-5
MFCD00060658
ZMPGXSFTXBOKFM-UHFFFAOYSA-N
1,3-dichloro-2-iodobenzene
140498
98%

Chemical Identifiers

19230-28-5
272.894
ZMPGXSFTXBOKFM-UHFFFAOYSA-N
140498
C1=CC(=C(C(=C1)Cl)I)Cl
C6H3Cl2I
MFCD00060658
#
1,3-dichloro-2-iodobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents