Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dichloro-2-fluorobenzene, 98%

$22.46 - $70.07

Chemical Identifiers

Chemical Name or Material 1,3-Dichloro-2-fluorobenzene
CAS 2268-05-5
Molecular Formula C6H3Cl2F
Formula Weight 165
MDL Number MFCD00010307
Beilstein 1862513
Synonym 2,6-Dichloro-1-fluorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2152603

  SDS
alfa aesar™
B2152603
1g Each for $22.46

AAB2152606

  SDS
alfa aesar™
B2152606
5g Each for $70.07
Description & Specifications

Specifications

Formula Weight 165
Assay 98%
MDL Number MFCD00010307
Beilstein 1862513
Boiling Point 168° to 169°C
Flash Point 73°C (163°F)
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 2268-05-5
Health Hazard 1 Warning
Synonym 2,6-Dichloro-1-fluorobenzene
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 1,3-Dichloro-2-fluorobenzene
Melting Point 36° to 40°C
Molecular Formula C6H3Cl2F
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.