Please login to your online account to display your discounted pricing

1,3-Dibromotetrafluorobenzene, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  336470050

 View more versions of this product

Catalog No. AC336470050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 307.87
Quantity 5g
Color Clear colorless to light yellow
TSCA TSCA
EINECS Number 216-326-3
CAS Max % 100.0
Health Hazard 1 LIGHT SENSITIVE
CAS Min % 97.5
Refractive Index 1.5160 to 1.5190 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Chemical Name or Material 1, 3-Dibromotetrafluorobenzene
MDL Number MFCD00031533
CAS 1559-87-1
Name Note 98%
Molecular Formula C6Br2F4
Assay Percent Range 98%

Specifications

Chemical Name or Material 1, 3-Dibromotetrafluorobenzene
Name Note 98%
CAS 1559-87-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6Br2F4
Formula Weight 307.87
MDL Number MFCD00031533
Accessories

LIGHT SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents