Please login to your online account to display your discounted pricing

1,3-Dibromotetrafluorobenzene, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 3-Dibromotetrafluorobenzene
Name Note 98%
CAS 1559-87-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6Br2F4
Formula Weight 307.87
MDL Number MFCD00031533
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC336470250

  SDS
thermo fisher scientific
336470250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC336470050

  SDS
thermo fisher scientific
336470050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 307.87
Quantity 25g
Color Clear colorless to light yellow
TSCA TSCA
EINECS Number 216-326-3
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
Refractive Index 1.5160 to 1.5190 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 1 LIGHT SENSITIVE
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98%
Molecular Formula C6Br2F4
Assay Percent Range 98%
Chemical Name or Material 1, 3-Dibromotetrafluorobenzene
MDL Number MFCD00031533
CAS 1559-87-1
Safety and Handling

LIGHT SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.