Learn More
1,3-Dibromopropane, 98%, stabilized, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 155040025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1,3-Dibromopropane | |
109-64-8 | |
100.0 | |
167.0°C | |
Authentic | |
Glass bottle | |
1.5215 to 1.5235 | |
MFCD00000255 | |
01, 110 | |
1.989 | |
trimethylene bromide, trimethylene dibromide, propane, 1,3-dibromo, alpha,gamma-dibromopropane, unii-yqr3048ix9, ccris 6711, .alpha.,.gamma.-dibromopropane, 1,3-dibromo propane, 1,3-dibromo-propane, .omega.,.omega.'-dibromopropane | |
VEFLKXRACNJHOV-UHFFFAOYSA-N | |
1,3-dibromopropane | |
400ppm butylene oxide | |
201.89 |
98% | |
97.5 | |
1.9890g/mL | |
54°C | |
97.5% min. (GC) | |
C3H6Br2 | |
Br(CH2)3Br | |
2.5L | |
13,313 | |
15, 9884 | |
Solubility in water: 1.7g/l (20°C). Other solubilities: soluble in organic solvents | |
BrCCCBr | |
201.89 | |
8001 | |
98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Flammable liquid and vapor.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Warning
EINECSNumber : 203-690-3
RTECSNumber : TX8575000
TSCA : TSCA
RUO â Research Use Only