missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dibromopropane, 98%, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 155041000

 View more versions of this product

Catalog No. AC155041000



Specifications

Specifications

1,3-Dibromopropane
54°C
Glass bottle
1.989
167.0°C
-34.0°C
98%
97.5
97.5% min. (GC)
Br(CH2)3Br
01, 110
15, 9884
Solubility in water: 1.7g/l (20°C). Other solubilities: soluble in organic solvents
C(CBr)CBr
201.89
201.89
98%
1.9890g/mL
Authentic
1.5215 to 1.5235
400ppm butylene oxide
Colorless to Yellow
100mL
109-64-8
100.0
C3H6Br2
MFCD00000255
13,313
.alpha.,.gamma.-dibromopropane
VEFLKXRACNJHOV-UHFFFAOYSA-N
1,3-dibromopropane
8001
Liquid

Chemical Identifiers

109-64-8
201.89
VEFLKXRACNJHOV-UHFFFAOYSA-N
8001
C(CBr)CBr
C3H6Br2
MFCD00000255
.alpha.,.gamma.-dibromopropane
1,3-dibromopropane
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Flammable liquid and vapor.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi

GHS Signal Word: Warning

EINECSNumber : 203-690-3

RTECSNumber : TX8575000

TSCA : TSCA

SDS Documents