missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dibromopropane, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar A146870I

 View more versions of this product

Catalog No. AAA146870I



Description

Description

Applications
1,3-Dibromopropane is used to prepare C3-bridged compounds through C-N coupling reactions. It is used in the preparation of 2,3,4,5-tetrahydro-7,7-diphenylimidazo- [2,1-b]-thiazine-6(7H)-one by reacting with potassium salt of 5,5-diphenyl-2-thiohydantoin.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

1,3-Dibromopropane
167°C
5000g
98%
Br(CH2)3Br
UN1993
14,9713
VEFLKXRACNJHOV-UHFFFAOYSA-N
1,3-dibromopropane
8001
Liquid
54°C (130°F)
-34°C
109-64-8
C3H6Br2
MFCD00000255
635662
trimethylene bromide, trimethylene dibromide, propane, 1,3-dibromo, alpha,gamma-dibromopropane, unii-yqr3048ix9, ccris 6711, .alpha.,.gamma.-dibromopropane, 1,3-dibromo propane, 1,3-dibromo-propane, .omega.,.omega.'-dibromopropane
C(CBr)CBr
201.889
201.9
98%

Chemical Identifiers

109-64-8
201.889
VEFLKXRACNJHOV-UHFFFAOYSA-N
8001
C(CBr)CBr
C3H6Br2
MFCD00000255
trimethylene bromide, trimethylene dibromide, propane, 1,3-dibromo, alpha,gamma-dibromopropane, unii-yqr3048ix9, ccris 6711, .alpha.,.gamma.-dibromopropane, 1,3-dibromo propane, 1,3-dibromo-propane, .omega.,.omega.'-dibromopropane
1,3-dibromopropane
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H315-H319
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

RTECSNumber : TX8575000

TSCA : TSCA

Documents