Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dibromoacetone, 98%

Chemical Identifiers

Chemical Name or Material 1,3-Dibromoacetone
CAS 816-39-7
Formula Weight 215.87
MDL Number MFCD00013540
UN Number UN1759
Beilstein 1740389
Synonym 1,3-Dibromo-2-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3732503

  SDS
alfa aesar™
H3732503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3732506

  SDS
alfa aesar™
H3732506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3732514

  SDS
alfa aesar™
H3732514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 29° to 30°C
Boiling Point 97° to 98°C (21mmHg)
Formula Weight 215.87
Chemical Name or Material 1,3-Dibromoacetone
TSCA TSCA
Physical Form Crystals or Fused Solid
Assay 98%
Color White to pale yellow
Quantity 1g
Sensitivity Moisture Sensitive
MDL Number MFCD00013540
CAS 816-39-7
UN Number UN1759
Beilstein 1740389
Synonym 1,3-Dibromo-2-propanone
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
DOT Information Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

Applications
1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.

Solubility
Soluble in acetone, chloroform, dichloromethane and methanol.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

2° to 8°C