missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dibromoacetone, 98%

Manufacturer:  Alfa Aesar™ H3732514

 View more versions of this product

Catalog No. AAH3732514



Description

Description

Applications
1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.

Solubility
Soluble in acetone, chloroform, dichloromethane and methanol.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
Specifications

Specifications

1,3-Dibromoacetone
97°C to 98°C (21mmHg)
29°C to 30°C
816-39-7
MFCD00013540
1740389
LQQKDSXCDXHLLF-UHFFFAOYSA-N
1,3-dibromopropan-2-one
69952
Crystals or Fused Solid
Moisture Sensitive
White to Yellow
25g
C3H4Br2O
UN1759
1,3-dibromoacetone, 1,3-dibromo-2-propanone, 2-propanone, 1,3-dibromo, 1,3-dibromopropanone, bromomethyl ketone, bromomethylketone, sgqdllrxbrwsp@, 1,3-dibromacetone, acmc-20ap4e, 2-propanone,3-dibromo
C(C(=O)CBr)Br
215.872
215.87
98%

Chemical Identifiers

816-39-7
215.872
LQQKDSXCDXHLLF-UHFFFAOYSA-N
69952
C(C(=O)CBr)Br
C3H4Br2O
MFCD00013540
1,3-dibromoacetone, 1,3-dibromo-2-propanone, 2-propanone, 1,3-dibromo, 1,3-dibromopropanone, bromomethyl ketone, bromomethylketone, sgqdllrxbrwsp@, 1,3-dibromacetone, acmc-20ap4e, 2-propanone,3-dibromo
1,3-dibromopropan-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

TSCA : TSCA

storageNote1 : 2° to 8°C

SDS Documents