missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dibromo-5-(trifluoromethoxy)benzene, 97%

$28.84 - $266.78

Chemical Identifiers

CAS 207226-31-1
Molecular Formula C7H3Br2F3O
Molecular Weight (g/mol) 319.903
MDL Number MFCD00075280
InChI Key UKHOUWWEEAOCTI-UHFFFAOYSA-N
Synonym 1,3-dibromo-5-trifluoromethoxy benzene, 1,3-dibromo-5-trifluoromethoxy-benzene, 3,5-dibromotrifluoromethoxybenzene, 3,5-dibromo trifluoromethoxy benzene, 3,5-dibromo-1-trifluoromethoxy benzene, 3,5-dibromo-alpha,alpha,alpha-trifluoroanisole, acmc-1cfab, ksc496c1r, 3,5-dibromo-triffluoromethoxybenzene, 3,5-dibromo-, ,-trifluoroanisole
PubChem CID 2761179
IUPAC Name 1,3-dibromo-5-(trifluoromethoxy)benzene
SMILES C1=C(C=C(C=C1Br)Br)OC(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6411506 View Documents Alfa Aesar™
H6411506
5g Each for $28.84
 
AAH6411514 View Documents Alfa Aesar™
H6411514
25g Each for $85.14
 
AAH6411522 View Documents Alfa Aesar™
H6411522
100g Each for $266.78
 
Specifications

Chemical Identifiers

207226-31-1
319.903
UKHOUWWEEAOCTI-UHFFFAOYSA-N
2761179
C1=C(C=C(C=C1Br)Br)OC(F)(F)F
C7H3Br2F3O
MFCD00075280
1,3-dibromo-5-trifluoromethoxy benzene, 1,3-dibromo-5-trifluoromethoxy-benzene, 3,5-dibromotrifluoromethoxybenzene, 3,5-dibromo trifluoromethoxy benzene, 3,5-dibromo-1-trifluoromethoxy benzene, 3,5-dibromo-alpha,alpha,alpha-trifluoroanisole, acmc-1cfab, ksc496c1r, 3,5-dibromo-triffluoromethoxybenzene, 3,5-dibromo-, ,-trifluoroanisole
1,3-dibromo-5-(trifluoromethoxy)benzene

Specifications

1.5080
Colorless
207226-31-1
MFCD00075280
UKHOUWWEEAOCTI-UHFFFAOYSA-N
1,3-dibromo-5-(trifluoromethoxy)benzene
2761179
Liquid
1,3-Dibromo-5-(trifluoromethoxy)benzene
83°C to 85°C (20mmHg)(literature)
5g
C7H3Br2F3O
1,3-dibromo-5-trifluoromethoxy benzene, 1,3-dibromo-5-trifluoromethoxy-benzene, 3,5-dibromotrifluoromethoxybenzene, 3,5-dibromo trifluoromethoxy benzene, 3,5-dibromo-1-trifluoromethoxy benzene, 3,5-dibromo-alpha,alpha,alpha-trifluoroanisole, acmc-1cfab, ksc496c1r, 3,5-dibromo-triffluoromethoxybenzene, 3,5-dibromo-, ,-trifluoroanisole
C1=C(C=C(C=C1Br)Br)OC(F)(F)F
319.903
319.9
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning