missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dibromo-5-(trifluoromethoxy)benzene, 97%

Manufacturer:  Alfa Aesar™ H6411522

 View more versions of this product

Catalog No. AAH6411522



Specifications

Specifications

1,3-Dibromo-5-(trifluoromethoxy)benzene
83°C to 85°C (20mmHg)(literature)
100g
C7H3Br2F3O
1,3-dibromo-5-trifluoromethoxy benzene, 1,3-dibromo-5-trifluoromethoxy-benzene, 3,5-dibromotrifluoromethoxybenzene, 3,5-dibromo trifluoromethoxy benzene, 3,5-dibromo-1-trifluoromethoxy benzene, 3,5-dibromo-alpha,alpha,alpha-trifluoroanisole, acmc-1cfab, ksc496c1r, 3,5-dibromo-triffluoromethoxybenzene, 3,5-dibromo-, ,-trifluoroanisole
C1=C(C=C(C=C1Br)Br)OC(F)(F)F
319.903
319.9
97%
1.5080
Colorless
207226-31-1
MFCD00075280
UKHOUWWEEAOCTI-UHFFFAOYSA-N
1,3-dibromo-5-(trifluoromethoxy)benzene
2761179
Liquid

Chemical Identifiers

207226-31-1
319.903
UKHOUWWEEAOCTI-UHFFFAOYSA-N
2761179
C1=C(C=C(C=C1Br)Br)OC(F)(F)F
C7H3Br2F3O
MFCD00075280
1,3-dibromo-5-trifluoromethoxy benzene, 1,3-dibromo-5-trifluoromethoxy-benzene, 3,5-dibromotrifluoromethoxybenzene, 3,5-dibromo trifluoromethoxy benzene, 3,5-dibromo-1-trifluoromethoxy benzene, 3,5-dibromo-alpha,alpha,alpha-trifluoroanisole, acmc-1cfab, ksc496c1r, 3,5-dibromo-triffluoromethoxybenzene, 3,5-dibromo-, ,-trifluoroanisole
1,3-dibromo-5-(trifluoromethoxy)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents