missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dibromo-5-fluorobenzene, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 263460250

 View more versions of this product

Catalog No. AC263460250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Liquid
Clear light yellow
100.0
Authentic
1.5760 to 1.5780 (20°C, 589nm)
Br2C6H3F
98%
MFCD00061119
253.9
25g
2.0180g/mL
Glass bottle
97.5
1435-51-4
C6H3Br2F
1, 3-Dibromo-5-fluorobenzene
98%

Chemical Identifiers

1435-51-4
100.0
C6H3Br2F
253.9
1, 3-Dibromo-5-fluorobenzene, 98%
97.5
98%
Br2C6H3F
MFCD00061119
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA