missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dibromo-5-fluorobenzene, 98%, ACROS Organics™

Chemical Identifiers

CAS 1435-51-4
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C6H3Br2F
Linear Formula Br2C6H3F
Formula Weight 253.9
MDL Number MFCD00061119
Chemical Name or Material 1, 3-Dibromo-5-fluorobenzene, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC263460250 SDS Thermo Fisher Scientific
263460250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1435-51-4
100.0
C6H3Br2F
253.9
1, 3-Dibromo-5-fluorobenzene, 98%
97.5
98%
Br2C6H3F
MFCD00061119
≥97.5%

Specifications

253.9
25g
2.0180g/mL
Glass bottle
97.5
1435-51-4
C6H3Br2F
MFCD00061119
≥97.5%
Liquid
Clear light yellow
100.0
Authentic
1.5760 to 1.5780 (20°C, 589nm)
Br2C6H3F
98%
1, 3-Dibromo-5-fluorobenzene, 98%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA