missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dibromo-5-fluorobenzene, 98+%

Manufacturer:  Alfa Aesar™ A1507806

 View more versions of this product

Catalog No. AAA1507806



Specifications

Specifications

1,3-Dibromo-5-fluorobenzene
204°C to 206°C
1435-51-4
MFCD00061119
3,5-dibromofluorobenzene, 3,5-dibromo-1-fluorobenzene, 1,3-dibromo-5-fluoro-benzene, benzene, 1,3-dibromo-5-fluoro, pubchem1052, acmc-1c2vf, intermediates-zcf02096, 3,5-bromo-1-fluorophenyl, 1,3dibromo-5-fluorobenzene, 3,5-dibromo-fluoro-benzene
C1=C(C=C(C=C1Br)Br)F
253.896
253.91
>110°C (230°F)
5g
C6H3Br2F
2353462
ASWYHZXKFSLNLN-UHFFFAOYSA-N
1,3-dibromo-5-fluorobenzene
137003
≥98%

Chemical Identifiers

1435-51-4
253.896
ASWYHZXKFSLNLN-UHFFFAOYSA-N
137003
C1=C(C=C(C=C1Br)Br)F
C6H3Br2F
MFCD00061119
3,5-dibromofluorobenzene, 3,5-dibromo-1-fluorobenzene, 1,3-dibromo-5-fluoro-benzene, benzene, 1,3-dibromo-5-fluoro, pubchem1052, acmc-1c2vf, intermediates-zcf02096, 3,5-bromo-1-fluorophenyl, 1,3dibromo-5-fluorobenzene, 3,5-dibromo-fluoro-benzene
1,3-dibromo-5-fluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents