Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dibromo-5-fluoro-2-iodobenzene, 97%

$32.09 - $32.09

Chemical Identifiers

Chemical Name or Material 1,3-Dibromo-5-fluoro-2-iodobenzene
CAS 62720-29-0
Molecular Formula C6H2Br2FI
Formula Weight 379.8
MDL Number MFCD00042232
Synonym 2,6-Dibromo-4-fluoroiodobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1557703

  SDS
alfa aesar™
A1557703
1g Each for $32.09
Description & Specifications

Specifications

Formula Weight 379.8
Melting Point 132° to 136°C
Chemical Name or Material 1,3-Dibromo-5-fluoro-2-iodobenzene
Assay 97%
Molecular Formula C6H2Br2FI
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
MDL Number MFCD00042232
Synonym 2,6-Dibromo-4-fluoroiodobenzene
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 62720-29-0
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.