missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dibromo-5-fluoro-2-iodobenzene, 97%

Manufacturer:  Alfa Aesar™ A1557703

 View more versions of this product

Catalog No. AAA1557703



Specifications

Specifications

1,3-Dibromo-5-fluoro-2-iodobenzene
1g
C6H2Br2FI
2,6-dibromo-4-fluoroiodobenzene, benzene, 1,3-dibromo-5-fluoro-2-iodo, 1,3-dibromo-5-fluoro-2-iodo-benzene, pubchem3449, acmc-1b2cd, ksc615o5d, 2.6-dibromo-4-fluoro iodobenzene, 2,6-dibromo-4-fluoro-1-iodobenzene, benzene,1,3-dibromo-5-fluoro-2-iodo
C1=C(C=C(C(=C1Br)I)Br)F
379.793
379.8
132°C to 136°C
62720-29-0
MFCD00042232
LIWKANDEJFABTQ-UHFFFAOYSA-N
1,3-dibromo-5-fluoro-2-iodobenzene
2724606
97%

Chemical Identifiers

62720-29-0
379.793
LIWKANDEJFABTQ-UHFFFAOYSA-N
2724606
C1=C(C=C(C(=C1Br)I)Br)F
C6H2Br2FI
MFCD00042232
2,6-dibromo-4-fluoroiodobenzene, benzene, 1,3-dibromo-5-fluoro-2-iodo, 1,3-dibromo-5-fluoro-2-iodo-benzene, pubchem3449, acmc-1b2cd, ksc615o5d, 2.6-dibromo-4-fluoro iodobenzene, 2,6-dibromo-4-fluoro-1-iodobenzene, benzene,1,3-dibromo-5-fluoro-2-iodo
1,3-dibromo-5-fluoro-2-iodobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents