Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dibromo-4-nitrobenzene, 98%

Chemical Identifiers

Chemical Name or Material 1,3-Dibromo-4-nitrobenzene
CAS 51686-78-3
Formula Weight 280.9
MDL Number MFCD02660416
UN Number UN3459
Synonym 2,4-Dibromo-1-nitrobenzene, 98%
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6173803

  SDS
alfa aesar™
H6173803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6173806

  SDS
alfa aesar™
H6173806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61738MD

  SDS
alfa aesar™
H61738MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 280.9
Chemical Name or Material 1,3-Dibromo-4-nitrobenzene
Physical Form Crystalline Powder
Assay 98%
MDL Number MFCD02660416
CAS 51686-78-3
UN Number UN3459
Synonym 2,4-Dibromo-1-nitrobenzene, 98%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITROBROMOBENZENES, SOLID
Color Yellow
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.