Learn More
1,3-Dibromo-2-propanol, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 406540250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 3-Dibromo-2-propanol | |
96-21-9 | |
100.0 | |
2.1300g/mL | |
46°C | |
94% min. (GC) | |
C3H6Br2O | |
CH2BrCHOHCH2Br | |
25 g | |
2.13 | |
Solubility in water: soluble. Other solubilities: very soluble in acetone,ethyl ether and ethanol | |
OC(CBr)CBr | |
217.89 | |
217.89 | |
Liquid |
pract., 85% | |
94.0 | |
Brown or Colorless to Yellow | |
219°C | |
Authentic | |
Glass bottle | |
1.5500 to 1.554 | |
MFCD00000216 | |
01, 365 | |
1,3-dibromo-2-propanol, 2-propanol, 1,3-dibromo, 1,3-dibromohydrin, glycerol 1,3-dibromohydrin, 2-hydroxy-1,3-dibromopropane, 1,3-dibromo-2-hydroxypropane, alpha-dibromohydrin, .alpha.-dibromohydrin, glycerol alpha,gamma-dibromohydrin, glycerol alpha,gamma-dibromohydrine | |
KIHQZLPHVZKELA-UHFFFAOYSA-N | |
1,3-dibromopropan-2-ol | |
7287 | |
95% |
Chemical Identifiers
96-21-9 | |
217.89 | |
KIHQZLPHVZKELA-UHFFFAOYSA-N | |
7287 | |
OC(CBr)CBr |
C3H6Br2O | |
MFCD00000216 | |
1,3-dibromo-2-propanol, 2-propanol, 1,3-dibromo, 1,3-dibromohydrin, glycerol 1,3-dibromohydrin, 2-hydroxy-1,3-dibromopropane, 1,3-dibromo-2-hydroxypropane, alpha-dibromohydrin, .alpha.-dibromohydrin, glycerol alpha,gamma-dibromohydrin, glycerol alpha,gamma-dibromohydrine | |
1,3-dibromopropan-2-ol |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Suspected of causing genetic defects.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN
GHS Signal Word: Warning
EINECSNumber : 202-489-8
RTECSNumber : UB0200000
TSCA : TSCA
RUO – Research Use Only