missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Dibromo-2,2-dimethoxypropane, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 35509-0050

 View more versions of this product

Catalog No. AC355090050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
261.94
White
100.0
Authentic
MFCD00051792
99%
22094-18-4
Crystalline Powder
5g
65° to 68°C
98.5
Glass bottle
1, 3-Dibromo-2, 2-dimethoxypropane
C5H10Br2O2

Chemical Identifiers

22094-18-4
100.0
C5H10Br2O2
MFCD00051792
≥98.5%
98.5
99%
261.94
1, 3-Dibromo-2, 2-dimethoxypropane

Safety and Handling

SDS & Safety Documents

TSCA : TSCA