Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dibromo-2,2-dimethoxypropane, 99%

$110.69 - $1,517.73

Chemical Identifiers

Chemical Name or Material 1,3-Dibromo-2,2-dimethoxypropane
CAS 22094-18-4
Molecular Formula C5H10Br2O2
Formula Weight 261.95
MDL Number MFCD00051792
Beilstein 1698992
Synonym 1,3-Dibromoacetone dimethyl acetal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1121706

 
alfa aesar™
A1121706
5g Each for $110.69

AAA1121722

 
alfa aesar™
A1121722
100g Each for $1,517.73

AAA1121714

 
alfa aesar™
A1121714
25g Each for $449.47
Description & Specifications

Specifications

Formula Weight 261.95
Chemical Name or Material 1,3-Dibromo-2,2-dimethoxypropane
Melting Point 64° to 67°C
Assay 99%
Quantity 5g
Molecular Formula C5H10Br2O2
MDL Number MFCD00051792
CAS 22094-18-4
Beilstein 1698992
Synonym 1,3-Dibromoacetone dimethyl acetal
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.