Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dibromo-2,2-diethylpropane, tech. 90%

$542.43 - $542.43

Chemical Identifiers

Chemical Name or Material 1,3-Dibromo-2,2-diethylpropane
CAS 67969-84-0
Purity Grade Technical Grade
Molecular Formula (CH3CH2)2C(CH2Br)2
Formula Weight 258.01
MDL Number MFCD00039190
Beilstein 1697848
Synonym 3,3-Bis(bromomethyl)pentane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0962918

  SDS
alfa aesar™
L0962918
50g Each for $542.43
Description & Specifications

Specifications

Formula Weight 258.01
MDL Number MFCD00039190
CAS 67969-84-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 1,3-Dibromo-2,2-diethylpropane
Assay 90%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Melting Point 38° to 41°C
Molecular Formula (CH3CH2)2C(CH2Br)2
Purity Grade Technical Grade
Quantity 50g
Synonym 3,3-Bis(bromomethyl)pentane
TSCA TSCA
Boiling Point 96° to 97°C (10mmHg)
Beilstein 1697848
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.