missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dibromo-2,2-diethylpropane, tech. 90%

Manufacturer:  Alfa Aesar™ L0962909

 View more versions of this product

Catalog No. AAL0962909



Specifications

Specifications

1,3-Dibromo-2,2-diethylpropane
38°C to 41°C
67969-84-0
MFCD00039190
3,3-bis bromomethyl pentane, 1,3-dibromo-2,2-diethylpropane, pentane, 3,3-bis bromomethyl, 3,3-bis bromomethyl pentane #, 2,2-diethyl-1,3-dibromopropane
CCC(CC)(CBr)CBr
257.997
258.01
Technical
96°C to 97°C (10mmHg)
10g
C7H14Br2
1697848
ZIQNZEFIQWBNHV-UHFFFAOYSA-N
3,3-bis(bromomethyl)pentane
106258
90%

Chemical Identifiers

67969-84-0
257.997
ZIQNZEFIQWBNHV-UHFFFAOYSA-N
106258
CCC(CC)(CBr)CBr
C7H14Br2
MFCD00039190
3,3-bis bromomethyl pentane, 1,3-dibromo-2,2-diethylpropane, pentane, 3,3-bis bromomethyl, 3,3-bis bromomethyl pentane #, 2,2-diethyl-1,3-dibromopropane
3,3-bis(bromomethyl)pentane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents