Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dibenzyl-5-cyanohexahydropyrimidine, 99%

Chemical Identifiers

Chemical Name or Material 1,3-Dibenzyl-5-cyanohexahydropyrimidine
CAS 86236-77-3
Molecular Formula C19H21N3
Formula Weight 291.39
MDL Number MFCD00051959
UN Number UN3439
Beilstein 5572924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0826406

  SDS
alfa aesar™
L0826406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0826414

  SDS
alfa aesar™
L0826414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 291.39
Chemical Name or Material 1,3-Dibenzyl-5-cyanohexahydropyrimidine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 86236-77-3
MDL Number MFCD00051959
Beilstein 5572924
Melting Point 77° to 82°C
Assay 99%
Quantity 5g
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
UN Number UN3439
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Molecular Formula C19H21N3
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.