missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Dibenzoyloxybenzene, 98%

Manufacturer:  Alfa Aesar™ A1198306

 View more versions of this product

Catalog No. AAA1198306



Specifications

Specifications

1,3-Dibenzoyloxybenzene
116°C to 119°C
5g
C20H14O4
2059467
SUQGLJRNDJRARS-UHFFFAOYSA-N
(3-benzoyloxyphenyl) benzoate
66742
94-01-9
98%
93°C (199°F)
MFCD00016576
Resorcinol dibenzoate
C1=CC=C(C=C1)C(=O)OC2=CC(=CC=C2)OC(=O)C3=CC=CC=C3
318.328
318.33

Chemical Identifiers

94-01-9
318.328
SUQGLJRNDJRARS-UHFFFAOYSA-N
66742
C1=CC=C(C=C1)C(=O)OC2=CC(=CC=C2)OC(=O)C3=CC=CC=C3
C20H14O4
MFCD00016576
Resorcinol dibenzoate
(3-benzoyloxyphenyl) benzoate
Safety and Handling

Safety and Handling

RTECSNumber : VH0590000

TSCA : TSCA

SDS Documents