missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Diaminopropane, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 112352500
Specifications
1, 3-Diaminopropane | |
48°C | |
Glass bottle | |
0.88 | |
140°C | |
-12°C | |
109-76-2,109-76-2 | |
100.0 | |
C3H10N2 | |
MFCD00008228 | |
13,160 | |
Solubility in water: soluble. Other solubilities: miscible with almost all common organic solvents | |
NCCCN | |
74.13 | |
CHEBI:16841 | |
Liquid |
0.8800g/mL | |
Authentic | |
1.4565 to 1.4585 | |
0.5% max. | |
Undesignated | |
250mL | |
98.5 | |
98.5% min. (GC) | |
H2N(CH2)3NH2 | |
04, 261 | |
1,3-Propanediamine, Trimethylenediamine | |
XFNJVJPLKCPIBV-UHFFFAOYSA-N | |
propane-1,3-diamine | |
5942 | |
74.13 | |
99% |
Chemical Identifiers
109-76-2 | |
74.13 | |
XFNJVJPLKCPIBV-UHFFFAOYSA-N | |
5942 | |
propane-1,3-diamine |
C3H10N2 | |
MFCD00008228 | |
1,3-Propanediamine, Trimethylenediamine | |
CHEBI:16841 | |
NCCCN |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Fatal in contact with skin.
Flammable liquid and vapor.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breat
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rin
GHS Signal Word: Danger
EINECSNumber : 203-702-7