missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Diaminopropane, 98%

Manufacturer:  Alfa Aesar™ A11932AE

 View more versions of this product

Catalog No. AAA11932AE


 

Description

Description

Applications
1,3-Diaminopropane is commonly used in the preparation of cholinesterase inhibitors and thrombosis inhibitors. It is also used as a polymer cross linkage agent, polyurethane extender and spray coatings. It is involved in the preparation of agrochemicals and pharmaceutical intermediates. Further, it acts as a building block in the synthesis of heterocyclic compounds, which finds application in textile finishing. In addition to this, it is used as a ligand and form coordination complexes.

Solubility
Miscible with water.

Notes
Hygroscopic. Air sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
Specifications

Specifications

1,3-Diaminopropane
134°C to 136°C
100mL
109-76-2
98%
MFCD00008228
605277
XFNJVJPLKCPIBV-UHFFFAOYSA-N
propane-1,3-diamine
428
74.13
48°C (118°F)
-12°C
>12
Amine-like
C3H10N2
UN2734
1,3-Propanediamine; Trimethylenediamine
C(CN)CN
74.127
CHEBI:15725

Chemical Identifiers

109-76-2
74.127
XFNJVJPLKCPIBV-UHFFFAOYSA-N
428
C3H10N2
MFCD00008228
1,3-Propanediamine; Trimethylenediamine
CHEBI:15725
Safety and Handling

Safety and Handling

GHS H Statement
H310-H314-H318-H226-H302
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.

RTECSNumber : TX6825000

TSCA : TSCA

SDS Documents