missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Diamino-2-hydroxypropane, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 112220250

 View more versions of this product

Catalog No. AC112220250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1, 3-Diamino-2-hydroxypropane
Authentic
114°C (5.0mmHg)
39°C to 45°C
95%
94.0
95%
H2NCH2CH(OH)CH2NH2
04, 290
Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,insoluble in diethyl ether,benzene
C(C(CN)O)N
90.12
90.12
95%
>110°C
Glass bottle
White to Yellow
25g
616-29-5
100.0
C3H10N2O
MFCD00008142
1,3 diamino-propan-2-ol, 1,3-diamino-2-hydroxy-propane, 1,3-diamino-2-hydroxypropane, 1,3-diamino-2-propanol, 2-hydroxy-1,3-diaminopropane, 2-hydroxy-1,3-propanediamine, 2-propanol, 1,3-diamino, 2-propanol,3-diamino, acmc-1b1xv, diaminoisopropanol
UYBWIEGTWASWSR-UHFFFAOYSA-N
1,3-diaminopropan-2-ol
61157
Low Melting Solid

Chemical Identifiers

616-29-5
90.12
UYBWIEGTWASWSR-UHFFFAOYSA-N
61157
C(C(CN)O)N
C3H10N2O
MFCD00008142
1,3 diamino-propan-2-ol, 1,3-diamino-2-hydroxy-propane, 1,3-diamino-2-hydroxypropane, 1,3-diamino-2-propanol, 2-hydroxy-1,3-diaminopropane, 2-hydroxy-1,3-propanediamine, 2-propanol, 1,3-diamino, 2-propanol,3-diamino, acmc-1b1xv, diaminoisopropanol
1,3-diaminopropan-2-ol
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-474-2

TSCA : TSCA

SDS Documents