Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Diacetylindole, 97%

$59.51 - $194.38

Chemical Identifiers

Chemical Name or Material 1,3-Diacetylindole
CAS 17537-64-3
Molecular Formula C12H11NO2
Formula Weight 201.23
MDL Number MFCD00039692
Beilstein 159285
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0369803

  SDS
alfa aesar™
L0369803
1g Each for $59.51

AAL0369806

  SDS
alfa aesar™
L0369806
5g Each for $194.38
Description & Specifications

Specifications

Formula Weight 201.23
Assay 97%
Molecular Formula C12H11NO2
MDL Number MFCD00039692
CAS 17537-64-3
Beilstein 159285
Melting Point 144° to 147°C
Quantity 1g
Chemical Name or Material 1,3-Diacetylindole