missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Diacetylbenzene, 99%, ACROS Organics™

Manufacturer:  Acros Organics 170090010

 View more versions of this product

Catalog No. AC170090010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1, 3-Diacetylbenzene
Authentic
1.5485 to 1.5505
Yellow
1g
6781-42-6
100.0
C10H10O2
MFCD00008740
VCHOFVSNWYPAEF-UHFFFAOYSA-N
1-(3-acetylphenyl)ethanone
23229
Liquid After Melting
>110°C
Glass bottle
150.0°C to 155.0°C (15.0mmHg)
35.0°C to 37.0°C
99%
98.5
99%
C6H4(COCH3)2
1,3-diacetylbenzene, 1,1'-1,3-phenylene diethanone, ethanone, 1,1'-1,3-phenylene bis, m-diacetylbenzene, benzene-1,3-bis acetyl, m-acetylacetophenone, m-acetyl acetophenone, 1-3-acetylphenyl ethanone, 1-3-acetylphenyl ethan-1-one, m-diacetyl benzene
CC(=O)C1=CC(=CC=C1)C(=O)C
162.188
162.19
99%

Chemical Identifiers

6781-42-6
162.188
VCHOFVSNWYPAEF-UHFFFAOYSA-N
23229
CC(=O)C1=CC(=CC=C1)C(=O)C
C10H10O2
MFCD00008740
1,3-diacetylbenzene, 1,1'-1,3-phenylene diethanone, ethanone, 1,1'-1,3-phenylene bis, m-diacetylbenzene, benzene-1,3-bis acetyl, m-acetylacetophenone, m-acetyl acetophenone, 1-3-acetylphenyl ethanone, 1-3-acetylphenyl ethan-1-one, m-diacetyl benzene
1-(3-acetylphenyl)ethanone
Safety and Handling

Safety and Handling

EINECSNumber : 229-842-9

TSCA : TSCA

SDS Documents