Please login to your online account to display your discounted pricing

1,3-Di-tert-butylbenzimidazolium chloride, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 3-Di-tert-butylbenzimidazolium chloride
CAS 946607-10-9
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C15H23ClN2
Formula Weight 266.81
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC435085000

  SDS
thermo fisher scientific
AC435085000
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC435080010

  SDS
thermo fisher scientific
AC435080010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Crystalline Powder
Formula Weight 266.81
Quantity 500mg
Color White to tan
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 1, 3-Di-tert-butylbenzimidazolium chloride
Elemental Analysis 64.15 to 70.90% C, 9.97 to 11.02% N
Assay Percent Range 96%
Molecular Formula C15H23ClN2
CAS 946607-10-9
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.