missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Di-tert-butylbenzimidazolium chloride, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC435085000

 View more versions of this product

Catalog No. AC435085000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1, 3-Di-tert-butylbenzimidazolium chloride
Authentic
204.0°C
946607-10-9
100.0
C15H23ClN2
CMQDFXSMZLANNA-UHFFFAOYSA-M
1,3-ditert-butylbenzimidazol-3-ium;chloride
16218265
Powder or Crystalline Powder
64.15 to 70.90% C, 9.97 to 11.02% N
Tan to White
500mg
95.0
96%
1,3-di-tert-butyl-1,3-benzodiazol-1-ium chloride, 1,3-di-tert-butyl-1h-benzimidazol-3-ium chloride, 1,3-di-tert-butyl-1h-benzo d imidazol-3-ium chloride, 1,3-di-tert-butylbenzimidazolium chloride
CC(C)(C)N1C=[N+](C2=CC=CC=C21)C(C)(C)C.[Cl-]
266.81
266.81
96%

Chemical Identifiers

946607-10-9
266.81
1,3-di-tert-butyl-1,3-benzodiazol-1-ium chloride, 1,3-di-tert-butyl-1h-benzimidazol-3-ium chloride, 1,3-di-tert-butyl-1h-benzo d imidazol-3-ium chloride, 1,3-di-tert-butylbenzimidazolium chloride
1,3-ditert-butylbenzimidazol-3-ium;chloride
C15H23ClN2
CMQDFXSMZLANNA-UHFFFAOYSA-M
16218265
CC(C)(C)N1C=[N+](C2=CC=CC=C21)C(C)(C)C.[Cl-]
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents