missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Di(p-tolyl)-1,1,3,3-tetramethyldisiloxane, 94%

$112.51 - $332.49

Chemical Identifiers

CAS 18055-70-4
Molecular Formula C18H26OSi2
Formula Weight 314.57
MDL Number MFCD09266163
Synonym 1,3-Bis(4-methylphenyl)-1,1,3,3-tetramethyldisiloxane
Chemical Name or Material 1,3-Di(p-tolyl)-1,1,3,3-tetramethyldisiloxane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2913003 SDS Alfa Aesar™
H2913003
1g Each for $112.51
AAH2913006 SDS Alfa Aesar™
H2913006
5g Each for $332.49
Specifications

Description

Applications
It is employed as a intermediates for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Description & Specifications

Chemical Identifiers

18055-70-4
314.57
1,3-Bis(4-methylphenyl)-1,1,3,3-tetramethyldisiloxane
C18H26OSi2
MFCD09266163
1,3-Di(p-tolyl)-1,1,3,3-tetramethyldisiloxane

Specifications

314.57
C18H26OSi2
1,3-Bis(4-methylphenyl)-1,1,3,3-tetramethyldisiloxane
1,3-Di(p-tolyl)-1,1,3,3-tetramethyldisiloxane
1g
18055-70-4
MFCD09266163
94%