Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Di-n-butylxanthine, 98%

$55.89 - $760.92

Chemical Identifiers

Chemical Name or Material 1,3-Di-n-butylxanthine
CAS 2850-36-4
Molecular Formula C13H20N4O2
Formula Weight 264.33
MDL Number MFCD00798591
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2049114

  SDS
alfa aesar™
B2049114
25g Each for $55.89

AAB2049122

  SDS
alfa aesar™
B2049122
100g Each for $179.31

AAB2049136

  SDS
alfa aesar™
B2049136
500g Each for $760.92
Description & Specifications

Specifications

Formula Weight 264.33
Quantity 25g
MDL Number MFCD00798591
CAS 2850-36-4
Chemical Name or Material 1,3-Di-n-butylxanthine
Assay 98%
Molecular Formula C13H20N4O2