Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Di(4-pyridyl)propane, 98%

Chemical Identifiers

Chemical Name or Material 1,3-Di(4-pyridyl)propane
CAS 17252-51-6
Molecular Formula C13H14N2
Formula Weight 198.27
MDL Number MFCD00038046
UN Number UN2811
Beilstein 150231
Synonym 4,4'-Trimethylenedipyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0242714

  SDS
alfa aesar™
L0242714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0242722

  SDS
alfa aesar™
L0242722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 198.27
Chemical Name or Material 1,3-Di(4-pyridyl)propane
MDL Number MFCD00038046
CAS 17252-51-6
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Molecular Formula C13H14N2
UN Number UN2811
Beilstein 150231
Synonym 4,4'-Trimethylenedipyridine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Flash Point 163°C (325°F)
Quantity 25g
TSCA TSCA
Assay 98%
Boiling Point 142°C (2mmHg)
Melting Point 53° to 56°C
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.