Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Di-2-thienyl-2-propen-1-one, 98+%

$67.58 - $218.58

Chemical Identifiers

Chemical Name or Material 1,3-Di-2-thienyl-2-propen-1-one
CAS 2309-48-0
Molecular Formula C11H8OS2
Formula Weight 220.31
MDL Number MFCD00014530
Beilstein 143294
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0085706

  SDS
alfa aesar™
L0085706
5g Each for $67.58

AAL0085714

  SDS
alfa aesar™
L0085714
25g Each for $218.58
Description & Specifications

Specifications

Formula Weight 220.31
Chemical Name or Material 1,3-Di-2-thienyl-2-propen-1-one
Assay ≥98%
CAS 2309-48-0
Molecular Formula C11H8OS2
MDL Number MFCD00014530
Beilstein 143294
Melting Point 98° to 100°C
Quantity 5g