missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Cyclooctadiene, 97%

Manufacturer:  Alfa Aesar™ L1051614

 View more versions of this product

Catalog No. AAL1051614



Specifications

Specifications

1,3-Cyclooctadiene
Characteristic
-53°C
1700-10-3
MFCD00001751
1901033
RRKODOZNUZCUBN-CCAGOZQPSA-N
(1Z,3Z)-cycloocta-1,3-diene
299882
97%
24°C (75°F)
142°C to 144°C
25g
C8H12
UN2520
1,3-cyclooctadiene, cyclooctadiene, 1z,3z-cycloocta-1,3-diene, cis,cis-1,3-cyclooctadiene, cycloocta-1,3-diene, 1,3-cyclooctadiene, z,z, cyclooctadienes, 1.3-cyclooctadiene, 1e,3z-1,3-cyclooctadiene, 1,3-cod
C1CCC=CC=CC1
108.184
108.18

Chemical Identifiers

1700-10-3
108.184
RRKODOZNUZCUBN-CCAGOZQPSA-N
299882
C1CCC=CC=CC1
C8H12
MFCD00001751
1,3-cyclooctadiene, cyclooctadiene, 1z,3z-cycloocta-1,3-diene, cis,cis-1,3-cyclooctadiene, cycloocta-1,3-diene, 1,3-cyclooctadiene, z,z, cyclooctadienes, 1.3-cyclooctadiene, 1e,3z-1,3-cyclooctadiene, 1,3-cod
(1Z,3Z)-cycloocta-1,3-diene
Safety and Handling

Safety and Handling

GHS H Statement
H304-H226-H315-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOOCTADIENES

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents