missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Cyclohexanedione, 97+%, may cont. up to 1% NaCl

Manufacturer:  Alfa Aesar™ A111090B

 View more versions of this product

Catalog No. AAA111090B


 

Description

Description

Applications
It is used as a nitisinone intermediate.

Solubility
Soluble in water (155 g/L) at 20°C, alcohol, chloroform, acetone, boiling benzene, ether (slightly), carbon disulfide (slightly), and petr ether (slightly).

Notes
Air & Light Sensitive. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
Specifications

Specifications

1,3-Cyclohexanedione
101°C to 105°C
504-02-9
C6H8O2
385888
Dihydroresorcinol
C1CC(=O)CC(=O)C1
112.128
CHEBI:17766
May contain up to 1% NaCl
1000g
≥97%
MFCD00001585
14,3178
HJSLFCCWAKVHIW-UHFFFAOYSA-N
cyclohexane-1,3-dione
10434
112.13

Chemical Identifiers

504-02-9
112.128
HJSLFCCWAKVHIW-UHFFFAOYSA-N
10434
C6H8O2
MFCD00001585
Dihydroresorcinol
CHEBI:17766
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents