missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Cyclohexanediol, Mixture of cis and trans 98%, ACROS Organics™

Manufacturer:  Acros Organics 111141000

 View more versions of this product

Catalog No. AC111141000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1, 3-Cyclohexanediol
Glass bottle
100g
504-01-8
100.0
C6H12O2
MFCD00039458
1,3-cyclohexanediol, resorcitol, 1,3-dihydroxycyclohexane, 1,3-benzenediol, hexahydro, cis-1,3-cyclohexandiol, trans-1,3-cyclohexanediol, trans-1,3-cyclohexanol, 1,3-cyclohexanediol,c&t, acmc-209ll9, acmc-209po7
C1CC(CC(C1)O)O
116.16
116.16
≥97.5%
Authentic
Yellow
98%
97.5
98%
C6H10(OH)2
06, 740
RLMGYIOTPQVQJR-UHFFFAOYSA-N
cyclohexane-1,3-diol
10433
Viscous Liquid After Melting

Chemical Identifiers

504-01-8
116.16
RLMGYIOTPQVQJR-UHFFFAOYSA-N
10433
C1CC(CC(C1)O)O
C6H12O2
MFCD00039458
1,3-cyclohexanediol, resorcitol, 1,3-dihydroxycyclohexane, 1,3-benzenediol, hexahydro, cis-1,3-cyclohexandiol, trans-1,3-cyclohexanediol, trans-1,3-cyclohexanol, 1,3-cyclohexanediol,c&t, acmc-209ll9, acmc-209po7
cyclohexane-1,3-diol
Safety and Handling

Safety and Handling

EINECSNumber : 207-979-5

TSCA : TSCA

SDS Documents