missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Cyclohexadiene, Stabilized 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 111100050

 View more versions of this product

Catalog No. AC111100050


 

Specifications Description & Specifications

Specifications

≥95%
Liquid
Clear colorless to light yellow
100.0
Authentic
592-57-4
96%
C6H8
0.1% BHT
80.13
5mL
95.0
Glass bottle
1, 3-Cyclohexadiene
96%
MFCD00001532
1.4730 to 1.4760 (20°C, 589nm)

Chemical Identifiers

592-57-4
100.0
C6H8
MFCD00001532
≥95%
95.0
96%
80.13
1, 3-Cyclohexadiene, 96%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-764-1

TSCA : TSCA