Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3-Chlorophenyl)ethanol, 97%

$64.34 - $233.55

Chemical Identifiers

Chemical Name or Material 1-(3-Chlorophenyl)ethanol
CAS 6939-95-3
Molecular Formula C8H9ClO
Formula Weight 156.61
MDL Number MFCD00021864
Beilstein 2042021
Synonym 3-Chloro-alpha-methylbenzyl alcohol; 3-Chlorophenyl methyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0478306

  SDS
alfa aesar™
L0478306
5g Each for $64.34

AAL0478314

  SDS
alfa aesar™
L0478314
25g Each for $233.55
Description & Specifications

Specifications

Formula Weight 156.61
Chemical Name or Material 1-(3-Chlorophenyl)ethanol
Molecular Formula C8H9ClO
Assay 97%
MDL Number MFCD00021864
CAS 6939-95-3
Beilstein 2042021
Synonym 3-Chloro-alpha-methylbenzyl alcohol; 3-Chlorophenyl methyl carbinol
Flash Point >110°C (230°F)
Boiling Point 83° to 85°C (1.5mmHg)
Quantity 5g